Learn More
Medchemexpress LLC HY-10984 500mg , Pomalidomide CAS:19171-19-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10984500MG
Encompass_Preferred
HY-10984 500mg Pomalidomide CAS: 19171-19-8 Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblon and induces degradation of essential Ikaros transcription factors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9932