Learn More
Cayman Chemical PomalIdomIde 5mg
SDP
Catalog No. 502982353
Encompass_Preferred
An inhibitor of cereblon (IC50 = ~3 µM for the human recombinant CRBN-DDB1 complex); inhibits autoubiquitination of CRBN in HEK293T cells expressing CRBN; inhibits proliferation of U266 myeloma cells at 0.1-10 µM; reduces tumor growth in a H929 R10-1 lenalidomide-resistant mouse xenograft model at 3 mg/kg per day when used in combination with dexamethasone
Catalog No. 50-298-2353 Supplier Cayman Chemical Supplier No. 198775 MG
An inhibitor of cereblon (IC50 = ~3 µM for the human recombinant CRBN-DDB1 complex); inhibits autoubiquitination of CRBN in HEK293T cells expressing CRBN; inhibits proliferation of U266 myeloma cells at 0.1-10 µM; reduces tumor growth in a H929 R10-1 lenalidomide-resistant mouse xenograft model at 3 mg/kg per day when used in combination with dexamethasone