Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-amido-C1-Br | 2351106-38-0 | MFCD32903493 | >98.0% | 394.18 g/mol | C15H12BrN3O5 | 5 MG
SDP

Catalog No. 5000263909
Encompass_Preferred

Pomalidomide-amido-C1-Br is a pomalidomide-derived E3 ligase ligand-linker conjugate used as a building block in targeted protein degradation and PROTAC research. Supplied as a solid research chemical, it is intended for in vitro studies and synthetic applications and should be stored under inert atmosphere at low temperature.

  • CAS number 2351106-38-0.
  • Appearance solid, white to light yellow.
  • Molecular formula C15H12BrN3O5.
  • Molecular weight 394.18 g/mol.
  • Purity >98.0%.
  • Storage under nitrogen at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Supplied as a 5 mg package.
  • SMILES O=C(NC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O)CBr.
  • Designed for use in PROTAC design and cereblon binding studies.

Catalog No. 50-002-63909 Supplier Medchemexpress LLC Supplier No. HY1306175MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Pomalidomide-amido-C1-Br is a pomalidomide-derived E3 ligase ligand-linker conjugate used as a building block in targeted protein degradation and PROTAC research. Supplied as a solid research chemical, it is intended for in vitro studies and synthetic applications and should be stored under inert atmosphere at low temperature.

  • CAS number 2351106-38-0.
  • Appearance solid, white to light yellow.
  • Molecular formula C15H12BrN3O5.
  • Molecular weight 394.18 g/mol.
  • Purity >98.0%.
  • Storage under nitrogen at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Supplied as a 5 mg package.
  • SMILES O=C(NC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O)CBr.
  • Designed for use in PROTAC design and cereblon binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.