Learn More
Medchemexpress LLC Pomalidomide-amido-c5-peg2-c6-chlorine | 1835705-69-5 | 98.75% | 594.10 | 25 MG
SDP
Catalog No. 5000347805
Encompass_Preferred
Pomalidomide-amido-C5-PEG2-C6-chlorine is a derivative of Pomalidomide. It is intended for research use only and is not sold to patients. The compound has a molecular weight of 594.10 and a chemical formula of C29H40ClN3O8. It appears as a white to off-white solid. This compound is cited in research related to the modular design of PROTACs for the degradation of oncogenic BCR-ABL.
  • It is a derivative of Pomalidomide.
  • It is used for research purposes only.
  • It is a solid with a white to off-white appearance.
  • It has been utilized in studies concerning PROTAC design for degrading oncogenic BCR-ABL.
Catalog No. 50-003-47805 Supplier Medchemexpress LLC Supplier No. 25MGHY137905
Pomalidomide-amido-C5-PEG2-C6-chlorine is a derivative of Pomalidomide. It is intended for research use only and is not sold to patients. The compound has a molecular weight of 594.10 and a chemical formula of C29H40ClN3O8. It appears as a white to off-white solid. This compound is cited in research related to the modular design of PROTACs for the degradation of oncogenic BCR-ABL.
  • It is a derivative of Pomalidomide.
  • It is used for research purposes only.
  • It is a solid with a white to off-white appearance.
  • It has been utilized in studies concerning PROTAC design for degrading oncogenic BCR-ABL.