Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-amido-C5-PEG2-C6-chlorine | 1835705-69-5 | 98.8% | 594.10 | 50 MG
SDP

Catalog No. 5000349649
Encompass_Preferred

Pomalidomide-amido-C5-PEG2-C6-chlorine is a derivative of Pomalidomide, intended for research use only. This product is supplied as a white to off-white solid.

  • High purity of 98.75%
  • Molecular weight of 594.10
  • Stable under recommended storage conditions
  • Store at 4°C under nitrogen
  • For research use only

Catalog No. 50-003-49649 Supplier Medchemexpress LLC Supplier No. HY13790550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-amido-C5-PEG2-C6-chlorine is a derivative of Pomalidomide, intended for research use only. This product is supplied as a white to off-white solid.

  • High purity of 98.75%
  • Molecular weight of 594.10
  • Stable under recommended storage conditions
  • Store at 4°C under nitrogen
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.