Learn More
Medchemexpress LLC Pomalidomide-amido-C5-PEG2-C6-chlorine | 1835705-69-5 | 99.7% | 594.10 g/mol | C29H40ClN3O8 | 5 MG
SDP
Catalog No. 5000222019
Encompass_Preferred
Pomalidomide-amido-C5-PEG2-C6-chlorine is a small-molecule derivative of pomalidomide provided for research use. It is supplied as a white to off-white solid intended for chemical biology and preclinical compound screening; not for human use.
  • High purity: 99.7%.
  • Molecular formula C29H40ClN3O8; molecular weight 594.10 g/mol.
  • Available in small package sizes: 1 mg, 5 mg, 10 mg, 25 mg, 50 mg.
  • White to off-white solid form.
  • Store powder at 4°C under nitrogen; in solvent, store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-22019 Supplier Medchemexpress LLC Supplier No. HY1379055MG
Pomalidomide-amido-C5-PEG2-C6-chlorine is a small-molecule derivative of pomalidomide provided for research use. It is supplied as a white to off-white solid intended for chemical biology and preclinical compound screening; not for human use.
  • High purity: 99.7%.
  • Molecular formula C29H40ClN3O8; molecular weight 594.10 g/mol.
  • Available in small package sizes: 1 mg, 5 mg, 10 mg, 25 mg, 50 mg.
  • White to off-white solid form.
  • Store powder at 4°C under nitrogen; in solvent, store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended for research use only; not for human or clinical use.