Learn More
Medchemexpress LLC Pomalidomide-amido-PEG3-C2-NH2 | 2328070-52-4 | 99.5% | 100 MG
SDP
Catalog No. 5000338639
Encompass_Preferred
Pomalidomide-amido-PEG3-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate designed for use in PROTAC technology. It incorporates a Pomalidomide-based cereblon ligand and a 3-unit PEG linker. This compound is for research use only and can be used to synthesize BI-3663, which degrades focal adhesion tyrosine kinase (PTK2).
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates pomalidomide-based cereblon ligand
  • Features a 3-unit PEG linker
  • Used in PROTAC technology
  • Can be used to synthesize BI-3663
  • Applicable in degrading PTK2 in hepatocellular carcinoma cell lines
Catalog No. 50-003-38639 Supplier Medchemexpress LLC Supplier No. HY130521100MG
Pomalidomide-amido-PEG3-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate designed for use in PROTAC technology. It incorporates a Pomalidomide-based cereblon ligand and a 3-unit PEG linker. This compound is for research use only and can be used to synthesize BI-3663, which degrades focal adhesion tyrosine kinase (PTK2).
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates pomalidomide-based cereblon ligand
  • Features a 3-unit PEG linker
  • Used in PROTAC technology
  • Can be used to synthesize BI-3663
  • Applicable in degrading PTK2 in hepatocellular carcinoma cell lines