Learn More
Medchemexpress LLC Pomalidomide-amido-PEG3-C2-NH2 | 2328070-52-4 | 99.55% | 50 MG
SDP
Catalog No. 5000339812
Encompass_Preferred
Pomalidomide-amido-PEG3-C2-NH2 is a high-purity, synthesized E3 ligase ligand-linker conjugate designed for research applications in PROTAC technology. This compound incorporates a Pomalidomide-based cereblon ligand and a 3-unit PEG linker. It can be utilized in the synthesis of BI-3663, a compound known to degrade focal adhesion tyrosine kinase (PTK2) in various cell lines.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Features a 3-unit PEG linker
  • Used in PROTAC technology
  • For research use only
  • Can be used to synthesize BI-3663
  • Degrades focal adhesion tyrosine kinase (PTK2)
  • Purity of 99.55%
  • Solid appearance, off-white to light yellow color
Catalog No. 50-003-39812 Supplier Medchemexpress LLC Supplier No. HY13052150MG
Pomalidomide-amido-PEG3-C2-NH2 is a high-purity, synthesized E3 ligase ligand-linker conjugate designed for research applications in PROTAC technology. This compound incorporates a Pomalidomide-based cereblon ligand and a 3-unit PEG linker. It can be utilized in the synthesis of BI-3663, a compound known to degrade focal adhesion tyrosine kinase (PTK2) in various cell lines.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Features a 3-unit PEG linker
  • Used in PROTAC technology
  • For research use only
  • Can be used to synthesize BI-3663
  • Degrades focal adhesion tyrosine kinase (PTK2)
  • Purity of 99.55%
  • Solid appearance, off-white to light yellow color