Learn More
Medchemexpress LLC Pomalidomide-amido-PEG3-C2-NH2 | 2328070-52-4 | 99.6% | 25 MG
SDP
Catalog No. 5000342448
Encompass_Preferred
Pomalidomide-amido-PEG3-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Pomalidomide-based cereblon ligand and a 3-unit PEG linker, components used in PROTAC technology. This product is intended for research use only and is not sold to patients.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide-based cereblon ligand
  • Uses a 3-unit PEG linker in PROTAC technology
  • Intended for research use only
  • Used in the synthesis of BI-3663
  • BI-3663 degrades focal adhesion tyrosine kinase (PTK2)
Catalog No. 50-003-42448 Supplier Medchemexpress LLC Supplier No. 25MGHY130521
Pomalidomide-amido-PEG3-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Pomalidomide-based cereblon ligand and a 3-unit PEG linker, components used in PROTAC technology. This product is intended for research use only and is not sold to patients.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide-based cereblon ligand
  • Uses a 3-unit PEG linker in PROTAC technology
  • Intended for research use only
  • Used in the synthesis of BI-3663
  • BI-3663 degrades focal adhesion tyrosine kinase (PTK2)