Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-amido-PEG3-C2-NH2 | 2328070-52-4 | 99.6% | 476.48 g·mol-1 | C22H28N4O8 | 5 MG
SDP

Catalog No. 5000263180
Encompass_Preferred

Pomalidomide-amido-PEG3-C2-NH2 is a pomalidomide-derived cereblon ligand-linker conjugate intended for PROTAC and chemical biology research. It couples a cereblon-binding motif to a three-unit PEG linker with a two-carbon spacer, offering a convenient chemical handle for attachment to target-binding ligands. The compound is supplied as an off-white to light yellow solid with high purity suitable for laboratory studies.

  • High purity (99.6% by HPLC).
  • Molecular weight 476.48 g·mol-1.
  • Molecular formula C22H28N4O8.
  • Physical form solid, off-white to light yellow.
  • Provides a PEG3 linker and C2 spacer for conjugation.
  • Designed for use as a cereblon ligand in PROTAC design.
  • Available in small research quantities such as 5 mg.

Catalog No. 50-002-63180 Supplier Medchemexpress LLC Supplier No. HY1305215MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-amido-PEG3-C2-NH2 is a pomalidomide-derived cereblon ligand-linker conjugate intended for PROTAC and chemical biology research. It couples a cereblon-binding motif to a three-unit PEG linker with a two-carbon spacer, offering a convenient chemical handle for attachment to target-binding ligands. The compound is supplied as an off-white to light yellow solid with high purity suitable for laboratory studies.

  • High purity (99.6% by HPLC).
  • Molecular weight 476.48 g·mol-1.
  • Molecular formula C22H28N4O8.
  • Physical form solid, off-white to light yellow.
  • Provides a PEG3 linker and C2 spacer for conjugation.
  • Designed for use as a cereblon ligand in PROTAC design.
  • Available in small research quantities such as 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.