Learn More
Medchemexpress LLC Pomalidomide-amido-PEG3-C2-NH2 | 2328070-52-4 | 99.6% | C22H28N4O8 | 10MG
SDP
Catalog No. 5000260171
Encompass_Preferred
Pomalidomide-amido-PEG3-C2-NH2 is a pomalidomide-derived cereblon ligand-linker conjugate intended for research use in PROTAC development and related E3 ligase conjugate chemistry. It offers reproducible properties for building probe molecules and degraders.
  • High purity (99.6%) for reliable assay and synthesis results.
  • Defined molecular weight (476.48 g/mol) and formula (C22H28N4O8) for accurate calculations.
  • Designed as a cereblon ligand module compatible with PROTAC assembly.
  • Provided with safety data sheet and product datasheet for lab use.
  • Available in multiple small-scale quantities to support screening and optimization.
Catalog No. 50-002-60171 Supplier Medchemexpress LLC Supplier No. HY13052110MG
Pomalidomide-amido-PEG3-C2-NH2 is a pomalidomide-derived cereblon ligand-linker conjugate intended for research use in PROTAC development and related E3 ligase conjugate chemistry. It offers reproducible properties for building probe molecules and degraders.
  • High purity (99.6%) for reliable assay and synthesis results.
  • Defined molecular weight (476.48 g/mol) and formula (C22H28N4O8) for accurate calculations.
  • Designed as a cereblon ligand module compatible with PROTAC assembly.
  • Provided with safety data sheet and product datasheet for lab use.
  • Available in multiple small-scale quantities to support screening and optimization.