Learn More
Medchemexpress LLC Pomalidomide-amino-PEG4-NH2 (hydrochloride) | 2331259-45-9 | 542.97 | 100 MG
SDP
Catalog No. 5000350449
Encompass_Preferred
Pomalidomide-amino-PEG4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Pomalidomide based cereblon ligand and a linker used in PROTAC technology. This compound is designed for use in PROTACs, which contain two different ligands connected by a linker. These PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Utilizes PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50449 Supplier Medchemexpress LLC Supplier No. HY138859A100MG
Pomalidomide-amino-PEG4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Pomalidomide based cereblon ligand and a linker used in PROTAC technology. This compound is designed for use in PROTACs, which contain two different ligands connected by a linker. These PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Utilizes PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins