Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-amino-PEG4-NH2 hydrochloride | 2331259-45-9 | >99.3% | 542.97 | 50 MG
SDP

Catalog No. 5000352020
Encompass_Preferred

Pomalidomide-amino-PEG4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. This compound incorporates the Pomalidomide-based cereblon ligand and a linker, commonly utilized in PROTAC technology. PROTACs, or Proteolysis-targeting chimeras, are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system through two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting the protein of interest.

  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Used in PROTAC technology
  • Target: E3 ligase ligand-linker conjugates
  • Pathway: PROTAC
  • Target: Cereblon
  • Soluble in DMSO (250 mg/mL)
  • Soluble in various in vivo solvent systems (≥ 6.25 mg/mL)

Catalog No. 50-003-52020 Supplier Medchemexpress LLC Supplier No. HY138859A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-amino-PEG4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. This compound incorporates the Pomalidomide-based cereblon ligand and a linker, commonly utilized in PROTAC technology. PROTACs, or Proteolysis-targeting chimeras, are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system through two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting the protein of interest.

  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Used in PROTAC technology
  • Target: E3 ligase ligand-linker conjugates
  • Pathway: PROTAC
  • Target: Cereblon
  • Soluble in DMSO (250 mg/mL)
  • Soluble in various in vivo solvent systems (≥ 6.25 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.