Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pomalidomide-Benzyl-Piperazine Hydrochloride

Catalog No. 92074650MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-074-650MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 92-074-650MG Supplier Sigma Aldrich Supplier No. 92074650MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protein degrader building block Pomalidomide-benzyl-piperazine hydrochloride enables the synthesis of molecules for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a Cereblon (CRBN)-recruiting ligand, a rigid linker, and a pendant amine for reactivity with a carboxylic acid on the target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a pendant amine, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.Technology Spotlight: Degrader Building Blocks for Targeted Protein DegradationPortal: Building PROTAC™ Degraders for Targeted Protein DegradationTargeted Protein Degradation by Small MoleculesSmall-Molecule PROTACS: New Approaches to Protein DegradationTargeted Protein Degradation: from Chemical Biology to Drug DiscoveryImpact of linker length on the activity of PROTACsPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 21160-83-8
Linear Formula C26H29N3O8
Quantity 50 mg
Synonym (2S,4R)-1-((S)-2-(3-((2-(2-Aminoethoxy)ethyl)(methyl)amino)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide; Crosslinker-E3 Ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader; VH032 conjugate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.