Learn More
Medchemexpress LLC Pomalidomide-C12-NH2 hydrochloride | 2862774-02-3 | 97.1% | 493.04 g/mol | C25H37ClN4O4 | 5 MG
SDP
Catalog No. 5000221408
Encompass_Preferred
Pomalidomide-C12-NH2 hydrochloride is an E3 ligase ligand-linker conjugate used as the Cereblon-directed ligand component in PROTAC design targeting KRAS G12C. Supplied as the hydrochloride salt for research use, it is provided as a solid in milligram scales and is characterized by high purity and defined storage recommendations.
Catalog No. 50-002-21408 Supplier Medchemexpress LLC Supplier No. HY1456155MG
Pomalidomide-C12-NH2 hydrochloride is an E3 ligase ligand-linker conjugate used as the Cereblon-directed ligand component in PROTAC design targeting KRAS G12C. Supplied as the hydrochloride salt for research use, it is provided as a solid in milligram scales and is characterized by high purity and defined storage recommendations.