Learn More
Medchemexpress LLC Pomalidomide-C12-NH2 hydrochloride | 2862774-02-3 | 97.1% | C25H37ClN4O4 | 100 MG
SDP
Catalog No. 5000356080
Encompass_Preferred
Pomalidomide-C12-NH2 hydrochloride is an E3 ligase ligand-linker conjugate of PROTAC KRAS G12C degrader-1 (HY-139186). This Cereblon-based KRASG12C degrader is intended for research use only and has not been fully validated for medical applications.
  • E3 ligase ligand-linker conjugate
  • Cereblon-based KRASG12C degrader
  • Suitable for laboratory research
  • Protect from light
  • Stable under recommended storage conditions
Catalog No. 50-003-56080 Supplier Medchemexpress LLC Supplier No. HY145615100MG
Pomalidomide-C12-NH2 hydrochloride is an E3 ligase ligand-linker conjugate of PROTAC KRAS G12C degrader-1 (HY-139186). This Cereblon-based KRASG12C degrader is intended for research use only and has not been fully validated for medical applications.
  • E3 ligase ligand-linker conjugate
  • Cereblon-based KRASG12C degrader
  • Suitable for laboratory research
  • Protect from light
  • Stable under recommended storage conditions