Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-C12-NH2 hydrochloride | 2862774-02-3 | 97.1% | C25H37ClN4O4 | 10MG
SDP

Catalog No. 5000259880
Encompass_Preferred

Pomalidomide-C12-NH2 hydrochloride is an E3 ligase ligand-linker conjugate that serves as the Cereblon-binding component in PROTAC constructs targeting KRAS G12C. Supplied as the hydrochloride salt, it is provided as a solid for use in targeted protein degradation research and biochemical assay development.

  • E3 ligase ligand-linker conjugate suitable for Cereblon-based PROTAC design.
  • Hydrochloride salt form; solid, light yellow to green yellow appearance.
  • High purity (97.1%).
  • Molecular formula C25H37ClN4O4; molecular weight 493.04 g·mol⁻1.
  • Recommended storage at -20°C; protect from light.
  • Available in small research sizes (5-500 mg).

Catalog No. 50-002-59880 Supplier Medchemexpress LLC Supplier No. HY14561510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-C12-NH2 hydrochloride is an E3 ligase ligand-linker conjugate that serves as the Cereblon-binding component in PROTAC constructs targeting KRAS G12C. Supplied as the hydrochloride salt, it is provided as a solid for use in targeted protein degradation research and biochemical assay development.

  • E3 ligase ligand-linker conjugate suitable for Cereblon-based PROTAC design.
  • Hydrochloride salt form; solid, light yellow to green yellow appearance.
  • High purity (97.1%).
  • Molecular formula C25H37ClN4O4; molecular weight 493.04 g·mol⁻1.
  • Recommended storage at -20°C; protect from light.
  • Available in small research sizes (5-500 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.