Learn More
Medchemexpress LLC Pomalidomide-C12-NH2 hydrochloride | 2862774-02-3 | 97.1% | C25H37ClN4O4 | 50 MG
SDP
Catalog No. 5000353366
Encompass_Preferred
Pomalidomide-C12-NH2 hydrochloride is an E3 ligase ligand-linker conjugate of PROTAC KRAS G12C degrader-1. This is a Cereblon-based KRASG12C degrader.
  • E3 ligase ligand-linker conjugate
  • Cereblon-based degrader
  • Intended for research use only
Catalog No. 50-003-53366 Supplier Medchemexpress LLC Supplier No. HY14561550MG
Pomalidomide-C12-NH2 hydrochloride is an E3 ligase ligand-linker conjugate of PROTAC KRAS G12C degrader-1. This is a Cereblon-based KRASG12C degrader.
  • E3 ligase ligand-linker conjugate
  • Cereblon-based degrader
  • Intended for research use only