Learn More
Medchemexpress LLC Pomalidomide-C2-amido-(C1-O-C5-O-C1)2-COOH | 2351103-63-2 | 93.9% | 648.7 | C31H44N4O11 | 5 MG
SDP
Catalog No. 5000264261
Encompass_Preferred
Pomalidomide-C2-amido-(C1-O-C5-O-C1)2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a pomalidomide-based cereblon ligand and a PEG-like linker; it is intended for use in PROTAC design, E3 ligase recruitment studies, and chemical probe synthesis.
  • Incorporates a pomalidomide-based cereblon ligand for CRBN recruitment.
  • Features a PEG-like C2 linker terminating in a carboxylic acid for conjugation.
  • Supplied as a purified small-molecule reagent for probe and PROTAC synthesis.
  • Available in multiple sizes to support screening and scale-up workflows.
  • Analytical data for purity and molecular properties provided for reproducibility.
Catalog No. 50-002-64261 Supplier Medchemexpress LLC Supplier No. HY1288485MG
Pomalidomide-C2-amido-(C1-O-C5-O-C1)2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a pomalidomide-based cereblon ligand and a PEG-like linker; it is intended for use in PROTAC design, E3 ligase recruitment studies, and chemical probe synthesis.
  • Incorporates a pomalidomide-based cereblon ligand for CRBN recruitment.
  • Features a PEG-like C2 linker terminating in a carboxylic acid for conjugation.
  • Supplied as a purified small-molecule reagent for probe and PROTAC synthesis.
  • Available in multiple sizes to support screening and scale-up workflows.
  • Analytical data for purity and molecular properties provided for reproducibility.