Learn More
Medchemexpress LLC HY-128846A 5mg Medchemexpress, Pomalidomide-C2-NH2 (hydrochloride) CAS:2305369-00-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128846A5MG
Encompass_Preferred
Medchemexpress, HY-128846A 5mg Pomalidomide-C2-NH2 (hydrochloride) CAS:2305369-00-8 Pomalidomide-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2636