Learn More
Medchemexpress LLC Pomalidomide-C3-NHS ester | 2828476-87-3 | 98.7% | 456.41 | 50 MG
SDP
Catalog No. 5000361585
Encompass_Preferred
Pomalidomide-C3-NHS ester is a synthesized E3 ligase ligand-linker conjugate, incorporating the Pomalidomide based cereblon ligand and NHS ester linker. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Includes NHS ester linker
  • Stable under recommended storage conditions
  • Ships at room temperature in continental US
  • Stores at 4°C, sealed, away from moisture
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-003-61585 Supplier Medchemexpress LLC Supplier No. HY14814250MG
Pomalidomide-C3-NHS ester is a synthesized E3 ligase ligand-linker conjugate, incorporating the Pomalidomide based cereblon ligand and NHS ester linker. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Includes NHS ester linker
  • Stable under recommended storage conditions
  • Ships at room temperature in continental US
  • Stores at 4°C, sealed, away from moisture
  • Soluble in DMSO at 100 mg/mL