Learn More
Medchemexpress LLC Pomalidomide-C3-NHS ester | 2828476-87-3 | 98.7% | 456.41 g·mol⁻1 | C21H20N4O8 | 10 MG
SDP
Catalog No. 5000286944
Encompass_Preferred
Pomalidomide-C3-NHS ester is a synthesized E3 ligase ligand-linker conjugate that combines a cereblon-binding pomalidomide scaffold with an NHS-activated C3 linker. It is designed as a conjugation-ready building block for targeted protein degradation chemistry and other bioconjugation applications, supplied with high purity and recommended storage conditions for research use.
  • E3 ligase recruiting building block for targeted protein degradation.
  • NHS ester functionalized C3 linker for efficient amide coupling to amines.
  • High purity (>98%) suitable for synthetic workflows.
  • Soluble in DMSO at 100 mg/mL for convenient stock preparation.
  • Offered in small research sizes, including 10 MG, for medicinal chemistry.
  • Store sealed, away from moisture, with recommended cold storage for solutions.
Catalog No. 50-002-86944 Supplier Medchemexpress LLC Supplier No. HY14814210MG
Pomalidomide-C3-NHS ester is a synthesized E3 ligase ligand-linker conjugate that combines a cereblon-binding pomalidomide scaffold with an NHS-activated C3 linker. It is designed as a conjugation-ready building block for targeted protein degradation chemistry and other bioconjugation applications, supplied with high purity and recommended storage conditions for research use.
  • E3 ligase recruiting building block for targeted protein degradation.
  • NHS ester functionalized C3 linker for efficient amide coupling to amines.
  • High purity (>98%) suitable for synthetic workflows.
  • Soluble in DMSO at 100 mg/mL for convenient stock preparation.
  • Offered in small research sizes, including 10 MG, for medicinal chemistry.
  • Store sealed, away from moisture, with recommended cold storage for solutions.