Learn More
Medchemexpress LLC Pomalidomide-C5-azide | 2434629-02-2 | 97.5% | 384.39 g·mol⁻¹ | C18H20N6O4 | 5 MG
SDP
Catalog No. 5000222331
Encompass_Preferred
Pomalidomide-C5-azide is a cereblon (CRBN) E3 ubiquitin ligase ligand-linker conjugate bearing a terminal azide for bioconjugation. Supplied as a light yellow solid with high reported purity, it is intended for research use and is commonly used as a building block in PROTAC synthesis and click-chemistry workflows.
  • Azide functional group enables click chemistry conjugation.
  • Serves as a cereblon (CRBN) ligand-linker conjugate for degrader synthesis.
  • High purity suitable for research applications.
  • Light yellow solid for convenient handling and storage.
  • Soluble in DMSO at high concentration (100 mg/mL) with ultrasonic assistance.
  • Intended for research use only; not for clinical use.
Catalog No. 50-002-22331 Supplier Medchemexpress LLC Supplier No. HY1393365MG
Pomalidomide-C5-azide is a cereblon (CRBN) E3 ubiquitin ligase ligand-linker conjugate bearing a terminal azide for bioconjugation. Supplied as a light yellow solid with high reported purity, it is intended for research use and is commonly used as a building block in PROTAC synthesis and click-chemistry workflows.
  • Azide functional group enables click chemistry conjugation.
  • Serves as a cereblon (CRBN) ligand-linker conjugate for degrader synthesis.
  • High purity suitable for research applications.
  • Light yellow solid for convenient handling and storage.
  • Soluble in DMSO at high concentration (100 mg/mL) with ultrasonic assistance.
  • Intended for research use only; not for clinical use.