Learn More
Medchemexpress LLC Pomalidomide-C5-azide | 2434629-02-2 | 97.5% | C18H20N6O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13933610MG
Encompass_Preferred
Pomalidomide-C5-azide is an E3 ubiquitin ligase cereblon (CRBN) ligand-linker conjugate bearing a terminal azide for bioorthogonal click chemistry. It is used as a synthetic building block for PROTACs and related chemical biology probes, enabling attachment of linkers or payloads via azide-alkyne cycloaddition. Intended for research use only.
  • Terminal azide suitable for copper-catalyzed and strain-promoted click reactions.
  • Cereblon-binding ligand enables E3 ligase recruitment in degrader constructs.
  • Useful building block for PROTAC synthesis and chemical biology probe development.
  • High purity (≈97.5%) appropriate for research applications.
  • Solid, light yellow appearance; store cold, protected from light, under inert gas when possible.
Catalog No. 50-002-60286