Learn More
Medchemexpress LLC Pomalidomide-C6-O-C5-O-C4-COOH | 2375774-54-0 | 98.55% | 559.65 | 25 MG
SDP
Catalog No. 5000362708
Encompass_Preferred
Pomalidomide-C6-O-C5-O-C4-COOH is an E3 ligase ligand-linker conjugate comprising a Pomalidomide-based cereblon ligand and linker. It can be used for the synthesis of PROTACs.
  • E3 ligase ligand-linker conjugate
  • Comprises a Pomalidomide-based cereblon ligand and linker
  • Suitable for the synthesis of PROTACs
Catalog No. 50-003-62708 Supplier Medchemexpress LLC Supplier No. 25MGHY148362
Pomalidomide-C6-O-C5-O-C4-COOH is an E3 ligase ligand-linker conjugate comprising a Pomalidomide-based cereblon ligand and linker. It can be used for the synthesis of PROTACs.
  • E3 ligase ligand-linker conjugate
  • Comprises a Pomalidomide-based cereblon ligand and linker
  • Suitable for the synthesis of PROTACs