Learn More
Medchemexpress LLC Pomalidomide-C6-O-C5-O-C4-COOH | 2375774-54-0 | 98.6% | 559.65 | 100 MG
SDP
Catalog No. 5000362876
Encompass_Preferred
Pomalidomide-C6-O-C5-O-C4-COOH is an E3 ligase ligand-linker conjugate, utilized in the synthesis of PROTACs. It comprises a Pomalidomide-based cereblon ligand and a linker.
  • E3 ligase ligand-linker conjugate
  • Comprises a cereblon ligand and a linker
  • Used for the synthesis of PROTACs
Catalog No. 50-003-62876 Supplier Medchemexpress LLC Supplier No. HY148362100MG
Pomalidomide-C6-O-C5-O-C4-COOH is an E3 ligase ligand-linker conjugate, utilized in the synthesis of PROTACs. It comprises a Pomalidomide-based cereblon ligand and a linker.
  • E3 ligase ligand-linker conjugate
  • Comprises a cereblon ligand and a linker
  • Used for the synthesis of PROTACs