Learn More
Medchemexpress LLC Pomalidomide-C6-O-C5-O-C4-COOH | 2375774-54-0 | 98.6% | 559.65 g·mol⁻¹ | C29H41N3O8 | 10 MG
SDP
Catalog No. 5000286235
Encompass_Preferred
Pomalidomide-C6-O-C5-O-C4-COOH is an E3 ligase ligand-linker conjugate that incorporates a pomalidomide-derived cereblon ligand attached to a carboxylated linker. It is intended as a building block for PROTAC synthesis and related protein degradation research. Store sealed, away from moisture, at -20°C; in solvent, store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Useful as an E3 ligase ligand-linker conjugate for PROTAC construction.
  • High purity suitable for synthetic applications (98.6%).
  • Molecular formula C29H41N3O8 and molecular weight 559.65 g·mol⁻¹.
  • Available in small, pre-weighed packages for medicinal chemistry workflows.
  • Chemical identification provided via CAS 2375774-54-0.
Catalog No. 50-002-86235 Supplier Medchemexpress LLC Supplier No. HY14836210MG
Add to Cart
Add to Cart
Pomalidomide-C6-O-C5-O-C4-COOH is an E3 ligase ligand-linker conjugate that incorporates a pomalidomide-derived cereblon ligand attached to a carboxylated linker. It is intended as a building block for PROTAC synthesis and related protein degradation research. Store sealed, away from moisture, at -20°C; in solvent, store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Useful as an E3 ligase ligand-linker conjugate for PROTAC construction.
  • High purity suitable for synthetic applications (98.6%).
  • Molecular formula C29H41N3O8 and molecular weight 559.65 g·mol⁻¹.
  • Available in small, pre-weighed packages for medicinal chemistry workflows.
  • Chemical identification provided via CAS 2375774-54-0.