Learn More
Apexbio Technology LLC Pomalidomide (CC-4047) 19171-19-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062531
Encompass_Preferred
Pomalidomide (CC-4047) an immunomodulatory derivative structurally related to Thalidomide exhibits anti-tumor and anti-angiogenic properties Chemically the molecule is distinguished by additional amino substituent at the fourth position of the phthalimide ring and two extra oxy groups Its mechanism of action includes modulation of specific cytokines within the tumor microenvironment (TNF- IL-6 IL-8 VEGF) suppression of tumor cell proliferation and modification of host immune cell activity Pomalidomide is widely studied in biomedical research particularly in therapeutic exploration for hematologic malignancies such as multiple myeloma including relapsed or refractory disease states
Catalog No. 50-000-62531 Supplier Apexbio Technology LLC Supplier No. A42125.1
Pomalidomide (CC-4047) an immunomodulatory derivative structurally related to Thalidomide exhibits anti-tumor and anti-angiogenic properties Chemically the molecule is distinguished by additional amino substituent at the fourth position of the phthalimide ring and two extra oxy groups Its mechanism of action includes modulation of specific cytokines within the tumor microenvironment (TNF- IL-6 IL-8 VEGF) suppression of tumor cell proliferation and modification of host immune cell activity Pomalidomide is widely studied in biomedical research particularly in therapeutic exploration for hematologic malignancies such as multiple myeloma including relapsed or refractory disease states