Learn More
Apexbio Technology LLC Pomalidomide (CC-4047) 19171-19-8 200mg
SDP
Catalog No. 5000061428
Encompass_Preferred
Pomalidomide (CC-4047) an immunomodulatory derivative structurally related to Thalidomide exhibits anti-tumor and anti-angiogenic properties Chemically the molecule is distinguished by additional amino substituent at the fourth position of the phthalimide ring and two extra oxy groups Its mechanism of action includes modulation of specific cytokines within the tumor microenvironment (TNF- IL-6 IL-8 VEGF) suppression of tumor cell proliferation and modification of host immune cell activity Pomalidomide is widely studied in biomedical research particularly in therapeutic exploration for hematologic malignancies such as multiple myeloma including relapsed or refractory disease states
Catalog No. 50-000-61428 Supplier Apexbio Technology LLC Supplier No. A4212200
Pomalidomide (CC-4047) an immunomodulatory derivative structurally related to Thalidomide exhibits anti-tumor and anti-angiogenic properties Chemically the molecule is distinguished by additional amino substituent at the fourth position of the phthalimide ring and two extra oxy groups Its mechanism of action includes modulation of specific cytokines within the tumor microenvironment (TNF- IL-6 IL-8 VEGF) suppression of tumor cell proliferation and modification of host immune cell activity Pomalidomide is widely studied in biomedical research particularly in therapeutic exploration for hematologic malignancies such as multiple myeloma including relapsed or refractory disease states