Learn More
Medchemexpress LLC Pomalidomide-coch2cl | 444287-84-7 | 97.0% | 349.73 g/mol | C15H12ClN3O5 | 250 MG
SDP
Catalog No. 5000285230
Encompass_Preferred
Pomalidomide-COCH2Cl is a synthetic derivative of pomalidomide bearing a chloroacetamide substituent on the benzyl ring. It is used as an E3 ligase ligand in PROTAC research and is supplied as a powder for laboratory research. Typical properties include molecular formula C15H12ClN3O5, molecular weight 349.73 g/mol, and a reported purity of 97.0%.
  • Derivative designed for E3 ligase binding in PROTAC applications.
  • Supplied as a stable powder suitable for long-term storage at low temperature.
  • Reported purity 97.0% to support analytical and synthetic workflows.
  • Molecular weight 349.73 g/mol and formula C15H12ClN3O5 for accurate formulation.
  • Certificate of analysis available for batch-specific quality data.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-85230 Supplier Medchemexpress LLC Supplier No. HYW391701250MG
Pomalidomide-COCH2Cl is a synthetic derivative of pomalidomide bearing a chloroacetamide substituent on the benzyl ring. It is used as an E3 ligase ligand in PROTAC research and is supplied as a powder for laboratory research. Typical properties include molecular formula C15H12ClN3O5, molecular weight 349.73 g/mol, and a reported purity of 97.0%.
  • Derivative designed for E3 ligase binding in PROTAC applications.
  • Supplied as a stable powder suitable for long-term storage at low temperature.
  • Reported purity 97.0% to support analytical and synthetic workflows.
  • Molecular weight 349.73 g/mol and formula C15H12ClN3O5 for accurate formulation.
  • Certificate of analysis available for batch-specific quality data.
  • Intended for research use only; not for human or clinical use.