Learn More
Medchemexpress LLC Pomalidomide-d3 | 2093128-28-8 | 99.0% | 276.27 | 50 MG
SDP
Catalog No. 5000344641
Encompass_Preferred
Pomalidomide-d3 is the deuterium labeled Pomalidomide, a third-generation immunomodulatory agent that acts as molecular glue. It interacts with the E3 ligase cereblon and induces the degradation of essential Ikaros transcription factors. It is for research use only and not sold to patients.
  • Deuterium labeled
  • Acts as a molecular glue
  • Interacts with E3 ligase cereblon
  • Induces degradation of essential Ikaros transcription factors
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-44641 Supplier Medchemexpress LLC Supplier No. HY10984S11MG
Pomalidomide-d3 is the deuterium labeled Pomalidomide, a third-generation immunomodulatory agent that acts as molecular glue. It interacts with the E3 ligase cereblon and induces the degradation of essential Ikaros transcription factors. It is for research use only and not sold to patients.
  • Deuterium labeled
  • Acts as a molecular glue
  • Interacts with E3 ligase cereblon
  • Induces degradation of essential Ikaros transcription factors
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs