Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-PEG2-acetic acid | 2143097-05-4 | 99.4% | 419.39 | 25 MG
SDP

Catalog No. 5000393926
Encompass_Preferred

Pomalidomide-PEG2-acetic acid is a conjugate composed of an E3 ligase ligand and a linker. It can be used for PROTAC synthesis.

  • Conjugate of an E3 ligase ligand and a linker
  • Can be used for PROTAC synthesis
  • Appearance: solid, light yellow to yellow

Catalog No. 50-003-93926 Supplier Medchemexpress LLC Supplier No. 25MGHY167759
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-PEG2-acetic acid is a conjugate composed of an E3 ligase ligand and a linker. It can be used for PROTAC synthesis.

  • Conjugate of an E3 ligase ligand and a linker
  • Can be used for PROTAC synthesis
  • Appearance: solid, light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.