Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pomalidomide-Peg2-Alkyne

Catalog No. 90152950MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-152-950MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-152-950MG Supplier Sigma Aldrich Supplier No. 90152950MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protein degrader builiding block Pomalidomide-PEG2-Alkyne enables the synthesis of molecules for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a Cereblon (CRBN)-recruiting ligand and a PEGylated crosslinker with pendant alkyne for click chemistry with an azide on the target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a pendant alkyne group, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.Technology Spotlight: Degrader Building Blocks for Targeted Protein DegradationPortal: Building PROTAC™ Degraders for Targeted Protein DegradationTargeted Protein Degradation by Small MoleculesSmall-Molecule PROTACS: New Approaches to Protein DegradationTargeted Protein Degradation: from Chemical Biology to Drug DiscoveryImpact of linker length on the activity of PROTACsPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 4392-87-4
Molecular Formula C12H7F3N2O3
Linear Formula C12H7F3N2O3
Quantity 25 mg
Synonym Carboxyl diazirine alkyne; Probe building block
Molecular Weight (g/mol) 284.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.