Learn More
Medchemexpress LLC Pomalidomide-PEG2-azide | 2267306-14-7 | 96.6% | C19H20N6O7 | 10MG
SDP
Catalog No. 5000261766
Encompass_Preferred
Pomalidomide-PEG2-azide is a pomalidomide-derived E3 ligase ligand bearing a two-unit polyethylene glycol spacer terminating in an azide group. It is provided as a solid research reagent intended for use as a ligand-linker building block in targeted protein degradation (PROTAC) synthesis and for bioorthogonal conjugation via click chemistry.
  • Provides cereblon-binding pomalidomide core for E3 ligase recruitment.
  • PEG2 spacer imparts flexibility and improves aqueous compatibility.
  • Terminal azide enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • High purity (96.6%) suitable for research applications.
  • Off-white to light yellow solid, highly soluble in DMSO.
  • Recommended storage at -20°C and protection from light.
Catalog No. 50-002-61766 Supplier Medchemexpress LLC Supplier No. HY13753710MG
Pomalidomide-PEG2-azide is a pomalidomide-derived E3 ligase ligand bearing a two-unit polyethylene glycol spacer terminating in an azide group. It is provided as a solid research reagent intended for use as a ligand-linker building block in targeted protein degradation (PROTAC) synthesis and for bioorthogonal conjugation via click chemistry.
  • Provides cereblon-binding pomalidomide core for E3 ligase recruitment.
  • PEG2 spacer imparts flexibility and improves aqueous compatibility.
  • Terminal azide enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • High purity (96.6%) suitable for research applications.
  • Off-white to light yellow solid, highly soluble in DMSO.
  • Recommended storage at -20°C and protection from light.