Learn More
Medchemexpress LLC Pomalidomide-PEG2-azide | 2267306-14-7 | MFCD32198604 | 96.6% | 444.40 g/mol | C19H20N6O7 | 5 MG
SDP
Catalog No. 5000222428
Encompass_Preferred
Pomalidomide-PEG2-azide is a pomalidomide-based cereblon (CRBN) ligand-linker conjugate featuring a two-unit polyethylene glycol spacer and an azide functional group. It is developed as a building block for targeted protein degrader (PROTAC) research and for modular conjugation via click chemistry, offering a balance of solubility and reactivity for medicinal chemistry applications.
  • Provides a pomalidomide scaffold for CRBN recruitment.
  • Two-unit PEG linker improves solubility and linker flexibility.
  • Azide handle enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Suitable for use as a PROTAC building block and linker module.
  • Available in small-scale quantities for medicinal chemistry and assay development.
Catalog No. 50-002-22428 Supplier Medchemexpress LLC Supplier No. HY1375375MG
Pomalidomide-PEG2-azide is a pomalidomide-based cereblon (CRBN) ligand-linker conjugate featuring a two-unit polyethylene glycol spacer and an azide functional group. It is developed as a building block for targeted protein degrader (PROTAC) research and for modular conjugation via click chemistry, offering a balance of solubility and reactivity for medicinal chemistry applications.
  • Provides a pomalidomide scaffold for CRBN recruitment.
  • Two-unit PEG linker improves solubility and linker flexibility.
  • Azide handle enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Suitable for use as a PROTAC building block and linker module.
  • Available in small-scale quantities for medicinal chemistry and assay development.