Learn More
Medchemexpress LLC Pomalidomide-PEG3-azide | 2267306-15-8 | MFCD32198606 | >98.0% | C21H24N6O8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13753810MG
Encompass_Preferred
Pomalidomide-PEG3-azide is an E3 ligase ligand-linker conjugate used as a building block in PROTAC synthesis and bioconjugation. It pairs a pomalidomide-derived cereblon-binding moiety with a three-unit polyethylene glycol linker terminated by an azide, enabling modular assembly of degrader molecules and efficient azide-alkyne click chemistry.
  • Azide functional group for click chemistry conjugation
  • PEG3 linker provides flexibility and solubility
  • Cereblon-binding ligand supports targeted protein degradation
  • High reported purity for reliable synthesis
  • Suitable for PROTAC assembly and chemical probe development
Catalog No. 50-002-59005