Learn More
Medchemexpress LLC Pomalidomide-PEG3-azide | 2267306-15-8 | MFCD32198606 | 98.9% | 488.45 | C21H24N6O8 | 5 MG
SDP
Catalog No. 5000223404
Encompass_Preferred
Pomalidomide-PEG3-azide is an E3 ligase ligand-linker conjugate that combines a pomalidomide-derived cereblon ligand with a three-unit polyethylene glycol linker terminated in an azide. It is intended for use in targeted protein degradation workflows and for click-chemistry functionalization to attach pomalidomide-based ligands to alkyne- or strained-alkyne-containing molecules.
  • Facilitates PROTAC construction via CRBN-mediated recruitment of E3 ligase.
  • Azide terminus enables copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • PEG3 linker provides flexible spacing between ligand and cargo.
  • High reported purity (≈98.9%) supports analytical and synthetic applications.
  • Soluble in DMSO at high concentration; store under inert atmosphere at -20°C to preserve stability.
Catalog No. 50-002-23404 Supplier Medchemexpress LLC Supplier No. HY1375385MG
Pomalidomide-PEG3-azide is an E3 ligase ligand-linker conjugate that combines a pomalidomide-derived cereblon ligand with a three-unit polyethylene glycol linker terminated in an azide. It is intended for use in targeted protein degradation workflows and for click-chemistry functionalization to attach pomalidomide-based ligands to alkyne- or strained-alkyne-containing molecules.
  • Facilitates PROTAC construction via CRBN-mediated recruitment of E3 ligase.
  • Azide terminus enables copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • PEG3 linker provides flexible spacing between ligand and cargo.
  • High reported purity (≈98.9%) supports analytical and synthetic applications.
  • Soluble in DMSO at high concentration; store under inert atmosphere at -20°C to preserve stability.