Learn More
Medchemexpress LLC HY-137538 50mg Medchemexpress, Pomalidomide-PEG3-azide CAS:2267306-15-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13753850MG
Encompass_Preferred
Medchemexpress, HY-137538 50mg Pomalidomide-PEG3-azide CAS:2267306-15-8 Pomalidomide-PEG3-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1498