Learn More
Medchemexpress LLC HY-128716A 100mg Medchemexpress, Pomalidomide-PEG3-C2-NH2 (TFA) CAS: Purity:>98%
SDP
Catalog No. 501932632
Encompass_Preferred
Medchemexpress, HY-128716A 100mg Pomalidomide-PEG3-C2-NH2 (TFA) CAS: Pomalidomide-PEG3-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2632 Supplier Medchemexpress LLC Supplier No. HY128716A100MG
Medchemexpress, HY-128716A 100mg Pomalidomide-PEG3-C2-NH2 (TFA) CAS: Pomalidomide-PEG3-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.