Learn More
Medchemexpress LLC Pomalidomide-PEG3-C2-NH2 hydrochloride | 2446474-09-3 | 96.6% | 250 MG
SDP
Catalog No. 5000341482
Encompass_Preferred
Pomalidomide-PEG3-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide-based cereblon ligand and a 3-unit PEG linker used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a Pomalidomide-based cereblon ligand
  • Includes a 3-unit PEG linker
  • Used in PROTAC technology
  • Appears as a light yellow to yellow solid
  • Structure consistent by 1H NMR spectrum
  • Structure consistent by LCMS
  • Purity of 96.6% by LCMS
  • For research use only
Catalog No. 50-003-41482 Supplier Medchemexpress LLC Supplier No. 250MGHY128716B
Pomalidomide-PEG3-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide-based cereblon ligand and a 3-unit PEG linker used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a Pomalidomide-based cereblon ligand
  • Includes a 3-unit PEG linker
  • Used in PROTAC technology
  • Appears as a light yellow to yellow solid
  • Structure consistent by 1H NMR spectrum
  • Structure consistent by LCMS
  • Purity of 96.6% by LCMS
  • For research use only