Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-PEG3-C2-NH2 TFA | 2414913-97-4 | 99.0% | 2 G
SDP

Catalog No. 5000342576
Encompass_Preferred

Pomalidomide-PEG3-C2-NH2 TFA is a synthetic E3 ligase ligand-linker conjugate, featuring a Pomalidomide-based cereblon ligand and a 3-unit PEG linker. This compound is designed for the synthesis of PROTACs and is intended for research use only.

  • Synthetic E3 ligase ligand-linker conjugate
  • Features a Pomalidomide-based cereblon ligand and 3-unit PEG linker
  • Utilized for the synthesis of PROTACs
  • High purity: 99.0%
  • Molecular weight: 562.49
  • Chemical formula: C23H29F3N4O9
  • Appears as a light yellow to yellow solid-liquid mixture
  • Soluble in DMSO, ethanol, and water
  • Offers multiple dissolution protocols for in vivo studies

Catalog No. 50-003-42576 Supplier Medchemexpress LLC Supplier No. HY128716A2G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-PEG3-C2-NH2 TFA is a synthetic E3 ligase ligand-linker conjugate, featuring a Pomalidomide-based cereblon ligand and a 3-unit PEG linker. This compound is designed for the synthesis of PROTACs and is intended for research use only.

  • Synthetic E3 ligase ligand-linker conjugate
  • Features a Pomalidomide-based cereblon ligand and 3-unit PEG linker
  • Utilized for the synthesis of PROTACs
  • High purity: 99.0%
  • Molecular weight: 562.49
  • Chemical formula: C23H29F3N4O9
  • Appears as a light yellow to yellow solid-liquid mixture
  • Soluble in DMSO, ethanol, and water
  • Offers multiple dissolution protocols for in vivo studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.