Learn More
Medchemexpress LLC Pomalidomide-PEG3-C2-NH2 TFA | 2414913-97-4 | 99.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY128716A50MG
Encompass_Preferred
Pomalidomide-PEG3-C2-NH2 TFA is a synthetic E3 ligase ligand-linker conjugate. It is composed of a Pomalidomide-based cereblon ligand and a 3-unit PEG linker. This compound is suitable for the synthesis of PROTACs. It has a molecular weight of 562.49, a chemical formula of C23H29F3N4O9, and typically appears as a light yellow to yellow solid-liquid mixture. This product is for research use only and not intended for human use.
  • Synthetic E3 ligase ligand-linker conjugate
  • Pomalidomide-based cereblon ligand
  • 3-unit PEG linker
  • Suitable for PROTACs synthesis
  • Target: Cereblon
Catalog No. 50-003-42053