Learn More
Medchemexpress LLC Pomalidomide-PEG3-C2-NH2 TFA (cereblon ligand-linker conjugates 5 TFA) | 2414913-97-4 | 99.0% | 500 MG
SDP
Catalog No. 5000341020
Encompass_Preferred
Pomalidomide-PEG3-C2-NH2 TFA is a synthetic E3 ligase ligand-linker conjugate. It is composed of a pomalidomide-based cereblon ligand and a 3-unit PEG linker.
  • Comprises a pomalidomide-based cereblon ligand.
  • Includes a 3-unit PEG linker.
  • Can be used for the synthesis of PROTACs.
Catalog No. 50-003-41020 Supplier Medchemexpress LLC Supplier No. HY128716A500MG
Pomalidomide-PEG3-C2-NH2 TFA is a synthetic E3 ligase ligand-linker conjugate. It is composed of a pomalidomide-based cereblon ligand and a 3-unit PEG linker.
  • Comprises a pomalidomide-based cereblon ligand.
  • Includes a 3-unit PEG linker.
  • Can be used for the synthesis of PROTACs.