Learn More
Medchemexpress LLC Pomalidomide-PEG4-azide | 2271036-47-4 | 99.8% | C23H30N6O8 | 250 MG
SDP
Catalog No. 5000352868
Encompass_Preferred
Pomalidomide-PEG4-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates a Pomalidomide-based cereblon ligand and a linker used in PROTAC technology. This click chemistry reagent contains an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. Pomalidomide-PEG4-azide is intended for research use only.
  • Incorporates a Pomalidomide-based cereblon ligand
  • Utilizes a linker used in PROTAC technology
  • Features an Azide group for click chemistry reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition
  • Facilitates strain-promoted alkyne-azide cycloaddition
  • Intended for research use only
Catalog No. 50-003-52868 Supplier Medchemexpress LLC Supplier No. 250MGHY141015
Pomalidomide-PEG4-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates a Pomalidomide-based cereblon ligand and a linker used in PROTAC technology. This click chemistry reagent contains an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. Pomalidomide-PEG4-azide is intended for research use only.
  • Incorporates a Pomalidomide-based cereblon ligand
  • Utilizes a linker used in PROTAC technology
  • Features an Azide group for click chemistry reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition
  • Facilitates strain-promoted alkyne-azide cycloaddition
  • Intended for research use only