Learn More
Medchemexpress LLC Pomalidomide-PEG4-C-COOH | 2097938-44-6 | 99.19% | 507.49 | 50 MG
SDP
Catalog No. 5000400818
Encompass_Preferred
Pomalidomide-PEG4-C-COOH (Cereblon Ligand -Linker Conjugate 1) is a synthesized E3 ligase ligand-linker conjugate that incorporates a Pomalidomide-based cereblon ligand and a 4-unit PEG linker, which are components used in PROTAC technology. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • A synthesized E3 ligase ligand-linker conjugate.
  • Incorporates Pomalidomide based cereblon ligand.
  • Includes a 4-unit PEG linker.
  • Used in PROTAC technology.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-004-00818 Supplier Medchemexpress LLC Supplier No. HY2193050MG
Pomalidomide-PEG4-C-COOH (Cereblon Ligand -Linker Conjugate 1) is a synthesized E3 ligase ligand-linker conjugate that incorporates a Pomalidomide-based cereblon ligand and a 4-unit PEG linker, which are components used in PROTAC technology. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • A synthesized E3 ligase ligand-linker conjugate.
  • Incorporates Pomalidomide based cereblon ligand.
  • Includes a 4-unit PEG linker.
  • Used in PROTAC technology.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.