Learn More
Medchemexpress LLC Pomalidomide-PEG4-C-COOH | 2097938-44-6 | 99.2% | 507.49 | 5 MG
SDP
Catalog No. 5000403200
Encompass_Preferred
Pomalidomide-PEG4-C-COOH (Cereblon Ligand -Linker Conjugate 1) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 4-unit PEG linker used in PROTAC technology. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Uses 4-unit PEG linker
  • Used in PROTAC technology
Catalog No. 50-004-03200 Supplier Medchemexpress LLC Supplier No. HY219305MG
Pomalidomide-PEG4-C-COOH (Cereblon Ligand -Linker Conjugate 1) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 4-unit PEG linker used in PROTAC technology. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Pomalidomide based cereblon ligand
  • Uses 4-unit PEG linker
  • Used in PROTAC technology