Learn More
Medchemexpress LLC Pomalidomide-PEG4-C2-NH2 | 2225940-52-1 | 95.1% | 492.52 | 50 MG
SDP
Catalog No. 5000437680
Encompass_Preferred
This synthesized E3 ligase ligand-linker conjugate incorporates a Pomalidomide-based cereblon ligand and a 4-unit PEG linker, utilized in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Features a 4-unit PEG linker
  • Used in PROTAC technology
Catalog No. 50-004-37680 Supplier Medchemexpress LLC Supplier No. HY11259950MG
This synthesized E3 ligase ligand-linker conjugate incorporates a Pomalidomide-based cereblon ligand and a 4-unit PEG linker, utilized in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Features a 4-unit PEG linker
  • Used in PROTAC technology