Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-PEG4-C2-NH2 | 2225940-52-1 | 95.11% | 492.52 | 2 G
SDP

Catalog No. 5000437315
Encompass_Preferred

Pomalidomide-PEG4-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker used in PROTAC technology. It is for research use only and not sold to patients.

  • Appearance: solid
  • Color: light yellow to yellow
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage: powder -20°C for 3 years, 4°C for 2 years. The compound is unstable in solutions, so freshly prepared solutions are recommended.
  • Target: cereblon
  • Related classifications: cancer, cancer targeted therapy, PROTAC E3 ligase ligand-linker conjugates.
  • Usage: for research use only. We do not sell to patients.

Catalog No. 50-004-37315 Supplier Medchemexpress LLC Supplier No. HY1125992G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-PEG4-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker used in PROTAC technology. It is for research use only and not sold to patients.

  • Appearance: solid
  • Color: light yellow to yellow
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage: powder -20°C for 3 years, 4°C for 2 years. The compound is unstable in solutions, so freshly prepared solutions are recommended.
  • Target: cereblon
  • Related classifications: cancer, cancer targeted therapy, PROTAC E3 ligase ligand-linker conjugates.
  • Usage: for research use only. We do not sell to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.