Learn More
Medchemexpress LLC HY-112599 500mg Medchemexpress, Pomalidomide-PEG4-C2-NH2 CAS:2225940-52-1 Purity:>98%
SDP
Catalog No. 501969109
Encompass_Preferred
Medchemexpress, HY-112599 500mg Pomalidomide-PEG4-C2-NH2 CAS:2225940-52-1 Pomalidomide-PEG4-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 4-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9109 Supplier Medchemexpress LLC Supplier No. HY112599500MG
Medchemexpress, HY-112599 500mg Pomalidomide-PEG4-C2-NH2 CAS:2225940-52-1 Pomalidomide-PEG4-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 4-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.