Learn More
Medchemexpress LLC Pomalidomide-peg4-c2-nh2 hydrochloride | 2357105-92-9 | 98.2% | 528.98 | 1 G
SDP
Catalog No. 5000437421
Encompass_Preferred
Pomalidomide-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker, used in the synthesis of PROTACs. It is for research use only and not sold to patients.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • These methods are for reference only.
Catalog No. 50-004-37421 Supplier Medchemexpress LLC Supplier No. HY112599B1G
Pomalidomide-PEG4-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a 4-unit PEG linker, used in the synthesis of PROTACs. It is for research use only and not sold to patients.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • These methods are for reference only.